ENA946832232 Enamine

tert-butyl 5-methoxy-1,2,3,6-tetrahydropyrazine-1-carboxylate

  • MDL number MFCD15142965
  • Cas Number : 590390-31-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form oil
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-177850

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-177850
Availability details for
Quantity
  • Availability
  • Add to Cart