ENA946832239 Enamine

N,N-dibenzylguanidine hydrochloride

  • MDL number MFCD01724226
  • Cas Number : 329-39-5
  • Molecular Weight : 275.78

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-366186

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-366186
Availability details for
Quantity
  • Availability
  • Add to Cart