ENA946832251 Enamine

N-(2-chloroethyl)carbamoyl chloride

  • MDL number MFCD19219721
  • Cas Number : 15872-02-3
  • Molecular Weight : 141.98

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. -10
Physical Form liquid
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-366223

Safety Information

signalword Danger
pcodes P234,P260,P264,P280,P301+P330+P331,P303+P361+P353,P304+P340,P305+P351+P338,P310,P363,P390,P405,P406,P501
MSDS https://www.enaminestore.com/catalog/EN300-366223
pictograms GHS05
hstatements H290,H314
Availability details for
Quantity
  • Availability
  • Add to Cart