ENA946832256 Enamine

3-(diethylamino)-1-phenylpropan-1-one

  • MDL number MFCD00071773
  • Cas Number : 94-38-2
  • Molecular Weight : 205.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-186867

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-186867
Availability details for
Quantity
  • Availability
  • Add to Cart