ENA946832260 Enamine

4-butoxy-3-(propan-2-yl)benzene-1-sulfonamide

  • MDL number MFCD11213165
  • Cas Number : 1094735-99-5
  • Molecular Weight : 271.38

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-366272

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-366272
Availability details for
Quantity
  • Availability
  • Add to Cart