ENA946832300 Enamine

2-({[(1,3-benzothiazol-2-yl)methyl](methyl)amino}methyl)aniline

  • MDL number MFCD30730604
  • Cas Number : 1913547-23-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-366392

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-366392
Availability details for
Quantity
  • Availability
  • Add to Cart