ENA946832304 Enamine

tert-butyl 3-(iodomethyl)-2-oxa-8-azaspiro[4.5]decane-8-carboxylate

  • MDL number MFCD15530221
  • Cas Number : 1250998-93-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form oil
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-366400

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-366400
Availability details for
Quantity
  • Availability
  • Add to Cart