ENA946832315 Enamine

ethyl 2-{[(tert-butoxy)carbonyl]amino}-3-hydroxy-3-phenylpropanoate

  • MDL number MFCD24534654
  • Cas Number : 1822543-61-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-366453

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-366453
Availability details for
Quantity
  • Availability
  • Add to Cart