ENA946832325 Enamine

3,5-bis(trifluoromethyl)pyrazin-2-amine

  • MDL number MFCD26521136
  • Cas Number : 1823898-82-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-366492

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-366492
Availability details for
Quantity
  • Availability
  • Add to Cart