ENA946832343 Enamine

(1R,2R)-4-oxocyclopentane-1,2-dicarboxylic acid

  • MDL number MFCD11559091
  • Cas Number : 67885-97-6
  • Molecular Weight : 172.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-366530

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-366530
Availability details for
Quantity
  • Availability
  • Add to Cart