ENA946832351 Enamine

methyl 2-(methanesulfonyloxy)acetate

  • MDL number MFCD28361357
  • Cas Number : 23550-26-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-366566

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-366566
Availability details for
Quantity
  • Availability
  • Add to Cart