ENA946832374 Enamine

methyl (2E)-4-chloro-4-oxobut-2-enoate

  • MDL number MFCD16876634
  • Cas Number : 17081-97-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 12-13
Purity 95
Storage temp. -10
Physical Form liquid
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-366689

Safety Information

signalword Danger
pcodes P234,P260,P261,P264,P270,P271,P280,P301+P312,P301+P330+P331,P303+P361+P353,P304+P340,P305+P351+P338,P310,P312,P330,P363,P390,P403+P233,P405,P406,P501
MSDS https://www.enaminestore.com/catalog/EN300-366689
pictograms GHS05,GHS07
hstatements H290,H302,H314,H335
Availability details for
Quantity
  • Availability
  • Add to Cart