ENA946832397 Enamine

1-(pyrazin-2-yl)propan-2-ol

  • MDL number MFCD08572185
  • Cas Number : 51460-71-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-212081

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-212081
Availability details for
Quantity
  • Availability
  • Add to Cart