ENA946832402 Enamine

6,7,8-trifluoro-1,4-dihydroquinolin-4-one

  • MDL number MFCD12192813
  • Cas Number : 1020087-32-4
  • Molecular Weight : 199.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-366768

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-366768
Availability details for
Quantity
  • Availability
  • Add to Cart