ENA946832404 Enamine

(4,4-dimethyloxolan-2-yl)methanesulfonyl chloride

  • MDL number MFCD30730401
  • Cas Number : 1934969-53-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-212355

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-212355
Availability details for
Quantity
  • Availability
  • Add to Cart