ENA946832408 Enamine

1-cyclohexylbutan-2-one

  • MDL number MFCD16819714
  • Cas Number : 24476-17-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-366777

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-366777
Availability details for
Quantity
  • Availability
  • Add to Cart