ENA946832446 Enamine

tert-butyl N-{[4-(trifluoromethyl)pyrrolidin-3-yl]methyl}carbamate

  • MDL number MFCD24545367
  • Cas Number : 168545-02-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form oil
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-366860

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-366860
Availability details for
Quantity
  • Availability
  • Add to Cart