ENA946832459 Enamine

ethyl 2-amino-5-ethoxybenzoate

  • MDL number MFCD12159975
  • Cas Number : 1178145-53-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-366897

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-366897
Availability details for
Quantity
  • Availability
  • Add to Cart