ENA946832461 Enamine

3-[(2-chloropyrimidin-4-yl)oxy]benzonitrile

  • MDL number MFCD01647389
  • Cas Number : 353259-11-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-366899

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-366899
Availability details for
Quantity
  • Availability
  • Add to Cart