ENA946833027 Enamine

6-[(tert-butoxy)carbonyl]-6-azabicyclo[3.2.1]octane-8-carboxylic acid

  • MDL number MFCD30730676
  • Cas Number : 2095409-23-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-366913

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-366913
Availability details for
Quantity
  • Availability
  • Add to Cart