ENA946833036 Enamine

[(2-bromophenyl)(phenyl)methyl](methyl)amine hydrochloride

  • MDL number MFCD30730686
  • Cas Number : 2094572-46-8
  • Molecular Weight : 312.64

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-366983

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-366983
Availability details for
Quantity
  • Availability
  • Add to Cart