ENA946833037 Enamine

lithium(1+) ion 2-(2,2-dimethylpropanamido)-4-methylpyridine-3-carboxylate

  • MDL number MFCD30730687
  • Cas Number : 2095411-12-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data Li+
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-366984

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-366984
Availability details for
Quantity
  • Availability
  • Add to Cart