ENA946833039 Enamine

2-methyl-7-(trifluoromethyl)-1,4-oxazepane

  • MDL number MFCD30730693
  • Cas Number : 2090998-14-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-367044

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-367044
Availability details for
Quantity
  • Availability
  • Add to Cart