ENA946833040 Enamine

5-(2,6-dichlorophenyl)-1,2,4-triazine-3-thiol

  • MDL number MFCD30730694
  • Cas Number : 2095410-60-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-367053

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-367053
Availability details for
Quantity
  • Availability
  • Add to Cart