ENA946833056 Enamine

5-(bromomethyl)-4-chloro-1,3-thiazole hydrobromide

  • MDL number MFCD30730711
  • Cas Number : 2095410-35-6
  • Molecular Weight : 293.41

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HBr
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-367204

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-367204
Availability details for
Quantity
  • Availability
  • Add to Cart