ENA946833059 Enamine

2,2,2-trifluoro-N-[(1,2,3,4-tetrahydroisoquinolin-3-yl)methyl]acetamide hydrochloride

  • MDL number MFCD30730715
  • Cas Number : 2094851-04-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-367227

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-367227
Availability details for
Quantity
  • Availability
  • Add to Cart