ENA946833063 Enamine

methyl 5-(hydroxymethyl)-2-methyl-1H-pyrrole-3-carboxylate

  • MDL number MFCD30730720
  • Cas Number : 2089213-09-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-367254

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-367254
Availability details for
Quantity
  • Availability
  • Add to Cart