ENA946833064 Enamine

potassium 1-(2-methoxyethyl)-1H-pyrazole-4-carboxylate

  • MDL number MFCD30730722
  • Cas Number : 2095409-72-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data K+
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-367259

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-367259
Availability details for
Quantity
  • Availability
  • Add to Cart