ENA946833071 Enamine

6-methoxy-5-(oxan-4-yl)-2,3,4,5-tetrahydropyridine

  • MDL number MFCD30730732
  • Cas Number : 2137787-66-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-367529

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-367529
Availability details for
Quantity
  • Availability
  • Add to Cart