ENA946833073 Enamine

2-(4,4-dimethyloxolan-2-yl)acetic acid

  • MDL number MFCD30730734
  • Cas Number : 2095410-08-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form oil
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-368358

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-368358
Availability details for
Quantity
  • Availability
  • Add to Cart