ENA946833076 Enamine

5-bromo-2-(1,3-dioxolan-2-yl)-4-methyl-1,3-thiazole

  • MDL number MFCD30730738
  • Cas Number : 2095410-24-3
  • Molecular Weight : 250.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-370466

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-370466
Availability details for
Quantity
  • Availability
  • Add to Cart