ENA946833079 Enamine

1-ethyl-5-(tributylstannyl)-1H-1,2,4-triazole

  • MDL number MFCD30730743

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-371055

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-371055
Availability details for
Quantity
  • Availability
  • Add to Cart