ENA946833611 Enamine

2-(cyclopentylmethyl)-2-methylpyrrolidine

  • MDL number MFCD21780436
  • Cas Number : 1499723-96-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. -10
Physical Form liquid
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-367192

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-367192
Availability details for
Quantity
  • Availability
  • Add to Cart