ENA946833613 Enamine

prop-2-en-1-yl 2,2-dimethylpropanoate

  • MDL number MFCD28361548
  • Cas Number : 15784-26-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-366921

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-366921
Availability details for
Quantity
  • Availability
  • Add to Cart