ENA946833619 Enamine

4-bromo-2-ethynylfuran

  • MDL number MFCD28657521
  • Cas Number : 1404051-34-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-366923

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-366923
Availability details for
Quantity
  • Availability
  • Add to Cart