ENA946833620 Enamine

2,2-dimethyl-3-phenylaziridine

  • MDL number MFCD19217123
  • Cas Number : 23040-89-3
  • Molecular Weight : 147.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-366925

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-366925
Availability details for
Quantity
  • Availability
  • Add to Cart