ENA946833643 Enamine

3-ethoxycyclohexan-1-ol

  • MDL number MFCD23735143
  • Cas Number : 51181-36-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-366908

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-366908
Availability details for
Quantity
  • Availability
  • Add to Cart