ENA946833645 Enamine

3-cyclopropanecarbonyloxane

  • MDL number MFCD21188414
  • Cas Number : 1472668-82-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-367223

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-367223
Availability details for
Quantity
  • Availability
  • Add to Cart