ENA946833651 Enamine

2,3-dihydrospiro[indene-1,3'-oxolane]-2',5'-dione

  • MDL number MFCD11134316
  • Cas Number : 72735-51-4
  • Molecular Weight : 202.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-367226

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-367226
Availability details for
Quantity
  • Availability
  • Add to Cart