ENA946833689 Enamine

4-methyl-1,4lambda5-azaphosphinan-4-one hydrochloride

  • MDL number MFCD30730725
  • Cas Number : 945459-80-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-367265

Safety Information

signalword Danger
pcodes P201,P202,P264,P270,P281,P301+P310,P308+P313,P330,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-367265
pictograms GHS06,GHS08
hstatements H301,H341,H351,H360FD
Availability details for
Quantity
  • Availability
  • Add to Cart