ENA946833721 Enamine

methyl 2-chloro-2-[4-(trifluoromethyl)phenyl]acetate

  • MDL number MFCD16723900
  • Cas Number : 1250104-84-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-367323

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-367323
Availability details for
Quantity
  • Availability
  • Add to Cart