ENA946833824 Enamine

5-methyl-4,5,6,7-tetrahydro-1H-indazole-4-carboxylic acid

  • MDL number MFCD30731242
  • Cas Number : 1546741-78-9
  • Molecular Weight : 180.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-372746

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-372746
Availability details for
Quantity
  • Availability
  • Add to Cart