ENA946833826 Enamine

(3S)-3-methyl-1,4-diazepan-2-one

  • MDL number MFCD19226104
  • Cas Number : 128427-25-8
  • Molecular Weight : 128.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-372748

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-372748
Availability details for
Quantity
  • Availability
  • Add to Cart