ENA946833842 Enamine

N-ethyl-1,3,4-oxadiazol-2-amine

  • MDL number MFCD30730735
  • Cas Number : 1851947-09-5
  • Molecular Weight : 113.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-368583

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-368583
Availability details for
Quantity
  • Availability
  • Add to Cart