ENA946833885 Enamine

2-(aminomethyl)-4-fluorophenol

  • MDL number MFCD12911482
  • Cas Number : 940874-99-7
  • Molecular Weight : 141.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-367043

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-367043
Availability details for
Quantity
  • Availability
  • Add to Cart