ENA946833890 Enamine

trimethyl({[4-(trimethylsilyl)but-3-yn-1-yl]oxy})silane

  • MDL number MFCD09868625
  • Cas Number : 58434-99-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-370279

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-370279
Availability details for
Quantity
  • Availability
  • Add to Cart