ENA946833901 Enamine

N-(2-cyano-4-nitrophenyl)acetamide

  • MDL number MFCD00115480
  • Cas Number : 73894-38-9
  • Molecular Weight : 205.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-370520

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-370520
Availability details for
Quantity
  • Availability
  • Add to Cart