ENA946833903 Enamine

2-(2-fluorophenyl)-N'-hydroxyethanimidamide

  • Cas Number : 872361-89-2
  • Molecular Weight : 168.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-370657

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-370657
Availability details for
Quantity
  • Availability
  • Add to Cart