ENA946833908 Enamine

6-methyl-4H,5H,6H,7H-pyrazolo[1,5-a]pyrimidine-3-carboxylic acid

  • MDL number MFCD29981520
  • Cas Number : 1545972-44-8
  • Molecular Weight : 181.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-370787

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-370787
Availability details for
Quantity
  • Availability
  • Add to Cart