ENA946833916 Enamine

4,5-dimethoxy-2,3-dihydro-1H-indole-2,3-dione

  • MDL number MFCD24562378
  • Cas Number : 76159-83-6
  • Molecular Weight : 207.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-367052

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-367052
Availability details for
Quantity
  • Availability
  • Add to Cart